missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-5-bromo-2-(1-pyrrolidinyl)pyrimidine, 95%, Thermo Scientific™

Catalog Number 15447705
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
885952-21-6
C15H16BrN3O
334.22
MFCD06798238
ZPBDALDYEJGXNL-UHFFFAOYSA-N
4-benzyloxy-5-bromo-2-pyrrolidin-1-yl pyrimidine, 4-benzyloxy-5-bromo-2-1-pyrrolidinyl pyrimidine, pyrimidine,5-bromo-4-phenylmethoxy-2-1-pyrrolidinyl
42553047
5-bromo-4-phenylmethoxy-2-pyrrolidin-1-ylpyrimidine
BrC1=CN=C(N=C1OCC1=CC=CC=C1)N1CCCC1
This item is not returnable. View return policy
885952-21-6
C15H16BrN3O
334.22
MFCD06798238
ZPBDALDYEJGXNL-UHFFFAOYSA-N
4-benzyloxy-5-bromo-2-pyrrolidin-1-yl pyrimidine, 4-benzyloxy-5-bromo-2-1-pyrrolidinyl pyrimidine, pyrimidine,5-bromo-4-phenylmethoxy-2-1-pyrrolidinyl
42553047
5-bromo-4-phenylmethoxy-2-pyrrolidin-1-ylpyrimidine
BrC1=CN=C(N=C1OCC1=CC=CC=C1)N1CCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 105°C to 108°C
Quantity 250 mg
Formula Weight 334.22
Percent Purity 95%
Chemical Name or Material 4-Benzyloxy-5-bromo-2-(1-pyrrolidinyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'