Få mere at vide
4-(Benzyloxy)-3-methoxy-benzyl Alcohol-d2, TRC
Artikelnummer. 30338197
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30338197 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338197 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28618750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Benzyloxy)-3-methoxy-benzyl Alcohol-d2
CAS 74719-60-1
Molekylær formel C15H14D2O3
Mængde 50 mg
Synonym (4-Benzyloxy-3-methoxyphenyl)methanol-d2, 3-Methoxy-4-(benzyloxy)benzyl Alcohol-d2, 4-(Benzyloxy)-3-methoxybenzyl Alcohol-d2, NSC 169518-d2, O-Benzylvanillyl Alcohol-d2, Vanillyl Alcohol Benzyl Ether-d2
InChI formel InChI=1S/C15H16O3/c1-17-15-9-13(10-16)7-8-14(15)18-11-12-5-3-2-4-6-12/h2-9,16H,10-11H2,1H3/i10D2
SMIL [2H]C([2H])(O)c1ccc(OCc2ccccc2)c(OC)c1
IUPAC navn dideuterio-(3-methoxy-4-phenylmethoxyphenyl)methanol
Molekylvægt (g/mol) 246.3
Formel vægt 246.1225