missing translation for 'onlineSavingsMsg'
Få mere at vide

4-(Benzyloxy)-3-methoxy-benzyl Alcohol-d2, TRC

Artikelnummer. 30330859
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30330859 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30330859 Leverandør Toronto Research Chemicals Leverandørnr. TRCB286187500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-(Benzyloxy)-3-methoxy-benzyl Alcohol-d2
CAS 74719-60-1
Molekylær formel C15H14D2O3
Mængde 500 mg
Synonym (4-Benzyloxy-3-methoxyphenyl)methanol-d2, 3-Methoxy-4-(benzyloxy)benzyl Alcohol-d2, 4-(Benzyloxy)-3-methoxybenzyl Alcohol-d2, NSC 169518-d2, O-Benzylvanillyl Alcohol-d2, Vanillyl Alcohol Benzyl Ether-d2
InChI formel InChI=1S/C15H16O3/c1-17-15-9-13(10-16)7-8-14(15)18-11-12-5-3-2-4-6-12/h2-9,16H,10-11H2,1H3/i10D2
SMIL [2H]C([2H])(O)c1ccc(OCc2ccccc2)c(OC)c1
IUPAC navn dideuterio-(3-methoxy-4-phenylmethoxyphenyl)methanol
Molekylvægt (g/mol) 246.3
Formel vægt 246.1225
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.