missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Benzylbenzaldehyde, 97%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Quantity | 1 g |
Sensitivity | Air sensitive |
Formula Weight | 196.24 |
Percent Purity | 97% |
Chemical Name or Material | 4-Benzylbenzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'