missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Benzoyl-4'-methyldiphenyl Sulfide, TRC

Artikelnummer. 30331919
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30331919 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30331919 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2082151G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-Benzoyl-4'-methyldiphenyl Sulfide
CAS 83846-85-9
Molekylær formel C20 H16 O S
Mængde 1 g
Synonym [4-[(4-Methylphenyl)thio]phenyl]phenylmethanone, 4-(4-Methylphenylthio)benzophenone, 4-(4-Tolylthio)benzophenone, 4-(p-Tolylthio)benzophenone, 4-Benzoyl-4'-methyldiphenyl sulfide, 4-Benzoyl-4'-methyldiphenyl thioether, BMS, Kayacure BMS, Lunacure BMS, Phenyl (p-tolylthio)phenyl ketone, Quantacure BMS, Runtecure 1030, SB-PI 705, Speedcure BMS
InChI formel InChI=1S/C20H16OS/c1-15-7-11-18(12-8-15)22-19-13-9-17(10-14-19)20(21)16-5-3-2-4-6-16/h2-14H,1H3
SMIL Cc1ccc(Sc2ccc(cc2)C(=O)c3ccccc3)cc1
IUPAC navn [4-(4-methylphenyl)sulfanylphenyl]-phenylmethanone
Molekylvægt (g/mol) 304.41
Formel vægt 304.0922
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.