Få mere at vide
4-Anilinomethylenepentenedioic Acid-d5 5-Methyl Ester, TRC
Artikelnummer. 30329600
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30329600 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329600 Leverandør Toronto Research Chemicals Leverandørnr. TRCA663752100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Anilinomethylenepentenedioic Acid-d5 5-Methyl Ester
CAS 1185237-80-2
Molekylær formel C13 D5 H8 N O4
Mængde 100 mg
Synonym 4-[(Phenylamino)methylene]-2-pentenedioic Acid 5-Methyl Ester
InChI formel InChI=1S/C13H13NO4/c1-18-13(17)10(7-8-12(15)16)9-14-11-5-3-2-4-6-11/h2-9,14H,1H3,(H,15,16)/b8-7+,10-9-/i2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(N\C=C(\C=C\C(=O)O)/C(=O)OC)c([2H])c1[2H]
IUPAC navn (2E,4Z)-4-methoxycarbonyl-5-(2,3,4,5,6-pentadeuterioanilino)penta-2,4-dienoic acid
Molekylvægt (g/mol) 252.277
Formel vægt 252.116