Få mere at vide
4-Anilino-4-oxobutanoic Acid-d5, TRC
Artikelnummer. 30339032
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30339032 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339032 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6639521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Anilino-d5-4-oxobutanoic Acid
CAS 840529-98-8
Molekylær formel C10 2H5 H6 N O3
Mængde 1 mg
Synonym Butanoic acid, 4-oxo-4-(phenyl-2,3,4,5,6-d5-amino)-, Butanoic acid, 4-oxo-4-(phenyl-d5-amino)- (9CI), 4-Oxo-4-(phenyl-2,3,4,5,6-d5-amino)butanoic acid, Succinanilic acid-d5, 4-Oxo-4-(phenylamino)butanoic acid D5 (phenyl-D5), 4-(Anilino-d5)-4-oxobutanoic acid, 4-Oxo-4-(N-phenyl-d5-amino)butanoic acid, N-(Phenyl-d5)butanedioic acid monoamide, N-(Phenyl-d5)succinamic acid, Succinic acid monoanilide-d5, Succinic monoanilide-d5
InChI formel InChI=1S/C10H11NO3/c12-9(6-7-10(13)14)11-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)(H,13,14)/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(NC(=O)CCC(=O)O)c([2H])c1[2H]
IUPAC navn 4-oxo-4-(2,3,4,5,6-pentadeuterioanilino)butanoic acid
Molekylvægt (g/mol) 198.23
Formel vægt 198.1053