missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Anilino-1-methylpiperidine, 98%, Thermo Scientific™

Catalog Number 11342319
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
22261-94-5
C12H18N2
190.29
MFCD06408133
GXJSWUKLJQHQFZ-UHFFFAOYSA-N
89642
1-methyl-N-phenylpiperidin-4-amine
CN1CCC(CC1)NC2=CC=CC=C2
This item is not returnable. View return policy
22261-94-5
C12H18N2
190.29
MFCD06408133
GXJSWUKLJQHQFZ-UHFFFAOYSA-N
89642
1-methyl-N-phenylpiperidin-4-amine
CN1CCC(CC1)NC2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 82°C to 86°C
Quantity 1 g
Synonym 4-anilino-1-methylpiperidine, n-1-methylpiperidin-4-yl aniline, 4-piperidinamine,1-methyl-n-phenyl, 1-methyl 4-piperidyl phenylamine, acmc-20akdg, 4-piperidinamine, 1-methyl-n-phenyl, 4-anilino-1-methylpiperidin, 4-anilino-1-methyl-piperidine, n-1-methyl-4-piperidinyl aniline, 1-methyl-n-phenyl-4-piperidinamine
Formula Weight 190.28
Percent Purity 98%
Chemical Name or Material 4-Anilino-1-methylpiperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'