Få mere at vide
4-Anilino-1-benzylpiperidine, TRC
Artikelnummer. 30136468
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136468 100 mg
30134106 50 mg
30009626 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136468 Leverandør Toronto Research Chemicals Leverandørnr. TRCA663680100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1155-56-2
Molekylær formel C18 H22 N2
Molekylvægt (g/mol) 266.38
Synonym 4-Piperidinamine, N-phenyl-1-(phenylmethyl)-, 4-anilino-1-benzyl- (7CI,8CI), N-Phenyl-1-(phenylmethyl)-4-piperidinamine, 1-Benzyl-4-(phenylamino)piperidine, 1-Benzyl-4-anilinopiperidine, 4-Anilino-1-benzylpiperidine, NSC 76613, 1-Benzyl-N-phenylpiperidin-4-amine
IUPAC navn 1-benzyl-N-phenylpiperidin-4-amine
SMIL C(N1CCC(CC1)Nc2ccccc2)c3ccccc3
Tekniske data
Kemisk navn eller materiale N-Phenyl-1-(phenylmethyl)-4-piperidinamine
Mængde 100 mg
InChI formel InChI=1S/C18H22N2/c1-3-7-16(8-4-1)15-20-13-11-18(12-14-20)19-17-9-5-2-6-10-17/h1-10,18-19H,11-15H2
Formel vægt 266.1783