Få mere at vide
4-Androsten-3-one-17beta-carboxylic Acid, Mikromol™
Artikelnummer. 17972516
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17972516 25 mg
17963496 100 mg
30463999 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17972516 Leverandør Mikromol Leverandørnr. MM1424.020025
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Androsten-3-one-17beta-carboxylic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Androsten-3-one-17beta-carboxylic Acid
CAS 302-97-6
Analyt- eller komponentnavne 4-Androsten-3-one-17beta-carboxylic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C20 H28 O3
Certificeringer/compliance ISO 17025
Synonym (17β)-3-Oxo-androst-4-ene-17-carboxylic Acid, Androst-4-ene-17β-carboxylic acid, 3-oxo- (6CI,7CI,8CI), 17β-Carboxyandrost-4-en-3-one, 3-Carbonyl-4-androstene-17β-carboxylic acid, 3-Oxoandrost-4-ene-17β-carboxylic acid, AD-Acid, AL 3793, CC 14837, Etienic acid, L 552.803, NSC 226121, SKF 106224, Testosterone-17β-carboxylic acid, 4-Androsten-3-one-17β-carboxylic Acid, 3-Oxo-4-androstene-17β-carboxylic Acid, SKF-106224, Dutasteride related
InChI formel InChI=1S/C20H28O3/c1-19-9-7-13(21)11-12(19)3-4-14-15-5-6-17(18(22)23)20(15,2)10-8-16(14)19/h11,14-17H,3-10H2,1-2H3,(H,22,23)/t14-,15-,16-,17+,19-,20-/m0/s1
SMIL C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)O
IUPAC navn (8S,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthrene-17-carboxylic acid
Molekylvægt (g/mol) 316.43
Mængde 25 mg
Formel vægt 316.2038 g/mol