Få mere at vide
4-Androsten-3,17-dione-2,2,4,6,6,16,16-d7, CDN
Artikelnummer. 30321104
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321104 0.05 g -
30314778 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321104 Leverandør CDN Leverandørnr. CDND53050.05G
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Androst-4-ene-3,17-dione-2,2,4,6,6,16,16-d7 (Major)
CAS 67034-85-9
Molekylær formel C19 D7 H19 O2
Mængde 0.05 g
InChI formel InChI=1 S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-16 H,3-10H2,1-2H3/t14-,15-,16-,18-,19-/m0/s1/i3D2,6D2,7D2,11 D
SMIL [2 H]C1=C2C([2 H])([2 H])C[C@@H]3 [C@H](CC[C@@]4(C)[C@H]3 CC([2 H])([2 H])C4=O)[C@@]2(C)CC([2 H])([2 H])C1=O
IUPAC navn (8 R,9 S,10 R,13 S,14 S)-2,2,4,6,6,16,16-heptadeuterio-10,13-dimethyl-1,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthrene-3,17-dione
Molekylvægt (g/mol) 293.4517
Formel vægt 293.2372 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity