Få mere at vide
4-Androsten-11beta-ol-3,17-dione-2,2,4,6,6,16,16-d7, CDN
Artikelnummer. 30327871
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327871 0.01 g
30319677 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327871 Leverandør CDN Leverandørnr. CDND61280.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 11 beta-Hydroxyandrost-4-ene-3,17-dione-2,2,4,6,6,16,16-D7
CAS 2331287-27-3
Molekylær formel C19 D7 H19 O3
Mængde 0.01 g
Synonym Androst-4-ene-3,17-dione-2,2,4,6,6,16,16-d7, 11-hydroxy-, (11β)- (ACI), (11β)-11-Hydroxyandrost-4-ene-3,17-dione-2,2,4,6,6,16,16-d7 (ACI), 11β-Hydroxyandrost-4-ene-3,17-dione-2,2,4,6,6,16,16-D7, 11β-Hydroxyandrost-4-ene-3,17-dione-2,2,4,6,6,16,16-d7, 4-Androsten-11β-ol-3,17-dione-2,2,4,6,6,16,16-d7
InChI formel InChI=1 S/C19H26O3/c1-18-8-7-12(20)9-11(18)3-4-13-14-5-6-16(22)19(14,2)10-15(21)17(13)18/h9,13-15,17,21 H,3-8,10H2,1-2H3/t13-,14-,15-,17+,18-,19-/m0/s1/i3D2,6D2,7D2,9 D
SMIL [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC([2 H])([2 H])C(=O)[C@@]4(C)C[C@H](O)[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O
IUPAC navn (8 S,9 S,10 R,11 S,13 S,14 S)-2,2,4,6,6,16,16-heptadeuterio-11-hydroxy-10,13-dimethyl-1,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthrene-3,17-dione
Molekylvægt (g/mol) 309.4511
Formel vægt 309.2321 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity