Få mere at vide
4-Aminopyridine-d6, CDN
Artikelnummer. 30311827
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311827 0.1 g
30320540 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311827 Leverandør CDN Leverandørnr. CDND51140.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dalfampridine-D6
CAS 45498-20-2
Molekylær formel C5 2H6 N2
Mængde 0.1 g
Synonym 4-Pyridin-2,3,5,6-d4-amine-d2 (9 CI), Dalfampridine-D6, Fampridine-D6, 4-Pyridylamine-d6
InChI formel InChI=1 S/C5H6N2/c6-5-1-3-7-4-2-5/h1-4 H,(H2,6,7)/i1D,2 D,3 D,4 D/hD2
SMIL [2 H]N([2 H])c1c([2 H])c([2 H])nc([2 H])c1[2 H]
IUPAC navn N,N,2,3,5,6-hexadeuteriopyridin-4-amine
Molekylvægt (g/mol) 100.15
Formel vægt 100.0908 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity