Få mere at vide
4-Aminophenol-13C6, TRC
Artikelnummer. 30134363
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134363 1 mg
30005550 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134363 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6189221MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246820-76-7
Molekylær formel 13C6H7NO
Molekylvægt (g/mol) 115.08
Synonym p-Aminophenol-3C6, 1-Amino-4-hydroxybenzene-3C6, 4-Amino-1-hydroxybenzene-3C6, 4-Hydroxy-1-aminobenzene-3C6, 4-Hydroxyaniline-3C6, 4-Hydroxybenzenamine-3C6, 4-Hydroxyphenylamine-3C6, Activol-3C6, Azol-3C6, Benzofur P-3C6, Certinal-3C6, Citol-3C6, Fouramine P-3C6, Fourrine 84-3C6, NSC 1545-3C6, Paranol-3C6, Rodinal-3C6, Unal-3C6, p-Hydroxyaniline-3C6, p-Hydroxyphenylamine-3C6;
IUPAC navn 4-amino(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-ol
SMIL [13CH]1=[13CH][13C](=[13CH][13CH]=[13C]1N)O
Tekniske data
Kemisk navn eller materiale 4-Aminophenol-13C6
Mængde 1 mg
InChI formel InChI=1S/C6H7NO/c7-5-1-3-6(8)4-2-5/h1-4,8H,7H2/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 115.0729