missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Aminophenazone-d3, TRC

Artikelnummer. 30324044
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324044 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324044 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6210022.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-Aminophenazone-d3
CAS 1329792-51-9
Molekylær formel C11H10D3N3O
Mængde 2.5 mg
Synonym 4-Amino-1,2-dihydro-1,5-(dimethyl-d3)-2-phenyl-3H-pyrazol-3-one, 4-Aminoantipyrine-d3, (2,3-Dihydro-1,5-(dimethyl-d3)-3-oxo-2-phenyl-1H-pyrazol-
InChI formel InChI=1S/C11H13N3O/c1-8-10(12)11(15)14(13(8)2)9-6-4-3-5-7-9/h3-7H,12H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])N1N(C(=O)C(=C1C)N)c2ccccc2
IUPAC navn 4-amino-5-methyl-2-phenyl-1-(trideuteriomethyl)pyrazol-3-one
Molekylvægt (g/mol) 206.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.