Få mere at vide
4-(Aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol, TRC
Artikelnummer. 30315765
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315765 10 mg
30314619 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30315765 Leverandør Toronto Research Chemicals Leverandørnr. TRCA47087010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyridoxamine
CAS 85-87-0
Molekylær formel C8 H12 N2 O2
Mængde 10 mg
Synonym Vitamin B6
InChI formel InChI=1S/C8H12N2O2/c1-5-8(12)7(2-9)6(4-11)3-10-5/h3,11-12H,2,4,9H2,1H3
SMIL Cc1ncc(CO)c(CN)c1O
IUPAC navn 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol
Molekylvægt (g/mol) 168.19
Formel vægt 168.0899