missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Aminomethyl-1-Boc-4-hydroxypiperidine, 97%

Brand:  Alfa Aesar™ H63079.MD

Additional Details : CAS Number : 392331-66-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15469568

  • 223.69 DKK / 250mg


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Description

Description

Applications
4-Aminomethyl-1-Boc-4-hydroxypiperidine can be used in agrochemical, pharmaceutical and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

392331-66-7
230.308
XYWCDAFPRBDRER-UHFFFAOYSA-N
23396319
CC(C)(C)OC(=O)N1CCC(CC1)(CN)O
C11H22N2O3
MFCD11579803
tert-butyl 4-aminomethyl-4-hydroxypiperidine-1-carboxylate, 1-boc-4-aminomethyl-4-hydroxypiperidine, tert-butyl 4-aminomethyl-4-hydroxytetrahydro-1 2h-pyridinecarboxylate, 1-boc-4-aminomethyl-4-hydroxypiperidine, 4-aminomethyl-1-boc-4-hydroxypiperidine, 4-aminomethyl-1-boc-piperidin-4-ol, tert-butyl 4-aminomethyl-4-hydroxypiperidinecarboxylate, tert-butyl 4-aminomethyl-4-hydroxy-piperidine-1-carboxylate, 4-aminomethyl-4-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl4-aminomethyl-4-hydroxypiperidine-1-carboxylate
tert-butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate

Specifications

4-Aminomethyl-1-Boc-4-hydroxypiperidine
White
392331-66-7
MFCD11579803
XYWCDAFPRBDRER-UHFFFAOYSA-N
tert-butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate
23396319
Solid
Air Sensitive
250mg
C11H22N2O3
tert-butyl 4-aminomethyl-4-hydroxypiperidine-1-carboxylate, 1-boc-4-aminomethyl-4-hydroxypiperidine, tert-butyl 4-aminomethyl-4-hydroxytetrahydro-1 2h-pyridinecarboxylate, 1-boc-4-aminomethyl-4-hydroxypiperidine, 4-aminomethyl-1-boc-4-hydroxypiperidine, 4-aminomethyl-1-boc-piperidin-4-ol, tert-butyl 4-aminomethyl-4-hydroxypiperidinecarboxylate, tert-butyl 4-aminomethyl-4-hydroxy-piperidine-1-carboxylate, 4-aminomethyl-4-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl4-aminomethyl-4-hydroxypiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)(CN)O
230.308
230.3
97%
Videos
SDS