Få mere at vide
4-Aminocyclohexanol-d10, TRC
Artikelnummer. 30315081
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30315081 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315081 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6039175MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Aminocyclohexanol-d10
CAS 1241045-80-6
Molekylær formel C6H3D10NO
Mængde 5 mg
Synonym 4-Hydroxycyclohexylamine-d10, 1-Amino-4-hydroxycyclohexane-d10, 4-Amino-1-cyclohexanol-d10
InChI formel InChI=1S/C6H13NO/c7-5-1-3-6(8)4-2-5/h5-6,8H,1-4,7H2/i1D2,2D2,3D2,4D2,5D,6D
SMIL NC1([2H])C([2H])([2H])C([2H])([2H])C(O)([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 4-aminocyclohexan-1,2,2,3,3,4,5,5,6,6-d10-1-ol
Molekylvægt (g/mol) 125.24
Formel vægt 125.16