Learn More
4-Aminobutyraldehyde diethyl acetal, 95%, ACROS Organics™
Brand:  Acros Organics 103260010
Additional Details : CAS Number : 6346-09-4 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Packaging Glass bottle
Quantity 1g
 View more versions of this product
Product Code. 10229803
  • 122.00 DKK / 1g
In stock, ready to ship: 5
Specifications
Chemical Identifiers
6346-09-4
161.24
GFLPSABXBDCMCN-UHFFFAOYSA-N
80672
CCOC(CCCN)OCC
C8H19NO2
MFCD00008227
4,4-diethoxybutylamine, 4-aminobutyraldehyde diethyl acetal, 1-butanamine, 4,4-diethoxy, 4,4-diethoxy-1-butanamine, ambda, 4,4-diethoxy-butylamine, acmc-2097jf, 4-aminobutanal diethyl acetal, ksc356g0p, 4,4-diethoxy-1-butanamine #
4,4-diethoxybutan-1-amine
Specifications
4-Aminobutyraldehyde diethyl acetal
62°C
Glass bottle
0.9
Colorless
6346-09-4
C8H19NO2
MFCD00008227
4,4-diethoxybutylamine, 4-aminobutyraldehyde diethyl acetal, 1-butanamine, 4,4-diethoxy, 4,4-diethoxy-1-butanamine, ambda, 4,4-diethoxy-butylamine, acmc-2097jf, 4-aminobutanal diethyl acetal, ksc356g0p, 4,4-diethoxy-1-butanamine #
GFLPSABXBDCMCN-UHFFFAOYSA-N
4,4-diethoxybutan-1-amine
80672
Liquid
0.9000g/mL
Authentic
1.4270 to 1.4290
196.0°C
1g
94% min. (GC)
H2N(CH2)3CH(OC2H5)2
04,319
Solubility in water: miscible.
CCOC(CCCN)OCC
161.24
161.24
95%
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 4-Aminobutyraldehyde diethyl acetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
SDS Documents