Få mere at vide
4-Aminobenzonitrile-2,3,5,6-d4, TRC
Artikelnummer. 30323816
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323816 10 mg
30315935 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323816 Leverandør Toronto Research Chemicals Leverandørnr. TRCA59171110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Aminobenzonitrile-2,3,5,6-D4
CAS 1331866-32-0
Molekylær formel C7 D4 H2 N2
Mængde 10 mg
Synonym Benzonitrile-2,3,5,6-d4, 4-amino- (ACI), 4-Aminobenzonitrile-2,3,5,6-d4 (ACI), 4-Aminobenzonitrile-2,3,5,6-D4, 4-Aminobenzonitrile-d4, 4-Cyanobenzeneamine-2,3,5,6-d4
InChI formel InChI=1S/C7H6N2/c8-5-6-1-3-7(9)4-2-6/h1-4H,9H2/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C#N)c([2H])c([2H])c1N
IUPAC navn 4-amino-2,3,5,6-tetradeuteriobenzonitrile
Molekylvægt (g/mol) 122.1606
Formel vægt 122.0782