Få mere at vide
4-Aminobenzoic Acid-d4, TRC
Artikelnummer. 30323962
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30323962 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323962 Leverandør Toronto Research Chemicals Leverandørnr. TRCA591502500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Aminobenzoic Acid-d4
CAS 350820-01-8
Molekylær formel C7 2H4 H3 N O2
Mængde 500 mg
Synonym Benzoic-2,3,5,6-d4 acid, 4-amino-, 4-Aminobenzoic-2,3,5,6-d4 acid, 4-Aminobenzoic-2,3,5,6-D4 acid, 4-Amino-2,3,5,6-tetradeuteriobenzoic acid
InChI formel InChI=1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C(=O)O)c([2H])c([2H])c1N
IUPAC navn 4-amino-2,3,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 141.16
Formel vægt 141.0728