Få mere at vide
4-Aminobenzamidine-d4 Dihydrochloride, TRC
Artikelnummer. 30315339
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30315339 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315339 Leverandør Toronto Research Chemicals Leverandørnr. TRCA58720210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Aminobenzamidine-d4 Dihydrochloride
CAS 1346600-28-9
Molekylær formel C7H7D4Cl2N3
Mængde 10 mg
Synonym 4-Aminobenzenecarboximidamide Dihydrochloride, 4-Amidinoaniline Dihydrochloride, 4-Aminobenzamidine Dihydrochloride, SC 67235, p-Aminobenzamidine Dihydrochloride
InChI formel InChI=1S/C7H9N3/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H3,9,10)/i1D,2D,3D,4D
SMIL NC1=C([2H])C([2H])=C(C(N)=N)C([2H])=C1[2H]
IUPAC navn 4-amino-2,3,5,6-tetradeuteriobenzenecarboximidamide
Molekylvægt (g/mol) 212.11