Få mere at vide
4-Aminoazobenzene, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Aminoazobenzene |
| CAS | 60-09-3 |
| Molekylær formel | C12 H11 N3 |
| Mængde | 500 mg |
| Synonym | Benzenamine, 4-(2-phenyldiazenyl)-, Benzenamine, 4-(phenylazo)- (9CI), C.I. Solvent Yellow 1 (7CI,8CI), 4-(2-Phenyldiazenyl)benzenamine, 4-(Phenylazo)aniline, 4-(Phenylazo)benzenamine, 4-(Phenyldiazenyl)aniline, 4-Aminoazobenzene, 4-Aminoazobenzol, Aniline yellow, Brasilazina Oil Yellow G, C.I. 11000, Cellitazol R, Ceres Yellow R, Fast Spirit Yellow, Fat Yellow AAB, NSC 2032, Oil Soluble Aniline Yellow, Oil Yellow AAB, Oil Yellow AB, Oil Yellow AN, Oil Yellow B, Organol Yellow 2A, Solvent Yellow 1, Solvent yellow, Somalia Yellow 2G, Sudan Yellow R, p-(Phenylazo)aniline, p-Aminoazobenzene, p-Aminoazobenzol, p-Aminodiphenylimide |
| InChI formel | InChI=1S/C12H11N3/c13-10-6-8-12(9-7-10)15-14-11-4-2-1-3-5-11/h1-9H,13H2 |
| SMIL | Nc1ccc(cc1)N=Nc2ccccc2 |
| IUPAC navn | 4-[(E)-phenyldiazenyl]aniline |
| Molekylvægt (g/mol) | 197.24 |
| Formel vægt | 197.0953 |