Få mere at vide
4-Aminoazobenzene-3,4'-disulfonic Acid Disodium Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Aminoazobenzene-3,4'-disulfonic Acid Disodium Salt |
| CAS | 2706-28-7 |
| Molekylær formel | C12 H9 N3 O6 S2 Na2 |
| Mængde | 250 mg |
| Synonym | Sodium 2-Amino-5-((4-sulfonatophenyl)diazenyl)benzenesulfonate, C.I. Acid Yellow 9, C.I. Acid Yellow 9, disodium Salt, Fast Yellow S, Acetyl Yellow G, Acid Yellow 9, Acid Yellow AT, Acid Yellow G, Acid Yellow G Kond, Acid Yellow Geigy, Acid yellow, Acilan Yellow Extra, Amacid Yellow RG, C Yellow 9, C.I. 13015, C.I. Food Yellow 2, Cilefa Yellow R, Disodium 4-aminoazobenzene-3,4'-disulfonate, E 105, E 105 (dye), Fast Yellow, Fast Yellow AB, Fast Yellow Extra, Fast Yellow Extra Specially Pure, Fast Yellow S Extra specially Pure, Fast Yellow SX Extra Specially Pure, Fast Yellow Y, Food Yellow 2, Golden Yellow RUAF, Hexacol Acid Yellow G, Kiton Yellow Extra, Lacquer Yellow T, Wool Yellow G, Yellow Fast, Yellow Acid |
| InChI formel | InChI=1S/C12H11N3O6S2.2Na/c13-11-6-3-9(7-12(11)23(19,20)21)15-14-8-1-4-10(5-2-8)22(16,17)18;;/h1-7H,13H2,(H,16,17,18)(H,19,20,21);;/q;2*+1/p-2 |
| SMIL | [Na+].[Na+].Nc1ccc(cc1S(=O)(=O)[O-])N=Nc2ccc(cc2)S(=O)(=O)[O-] |
| IUPAC navn | disodium;2-amino-5-[(4-sulfonatophenyl)diazenyl]benzenesulfonate |
| Molekylvægt (g/mol) | 401.32 |
| Formel vægt | 400.9728 |