Få mere at vide
4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-iodo-2-methoxy-benzamide, TRC
Artikelnummer. 30009714
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009714 25 mg
30006663 5 mg
30007407 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009714 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60941025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 176849-91-5
Molekylær formel C15 H22 I N3 O2
Molekylvægt (g/mol) 403.26
Synonym 4-Amino-N-[[(2RS)-1-ethyl-pyrrolidin-2-yl]methyl]-5-iodo-2-methoxy-benzamide, Amisulpride Imp. C (EP)
IUPAC navn 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-iodo-2-methoxybenzamide
SMIL CCN1CCCC1CNC(=O)c2cc(I)c(N)cc2OC
Tekniske data
Kemisk navn eller materiale 4-Amino-N-[((2RS)-1-ethyl-pyrrolidin-2-yl)methyl]-5-iodo-2-methoxy-benzamide
Analyt- eller komponentnavne 4-Amino-N-[((2RS)-1-ethyl-pyrrolidin-2-yl)methyl]-5-iodo-2-methoxy-benzamide
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C15H22IN3O2/c1-3-19-6-4-5-10(19)9-18-15(20)11-7-12(16)13(17)8-14(11)21-2/h7-8,10H,3-6,9,17H2,1-2H3,(H,18,20)
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 25 mg
Formel vægt 403.0757