Få mere at vide
4-Amino-D,L-benzylsuccinic Acid, TRC
Artikelnummer. 30313424
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313424 25 mg
30311069 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30313424 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60150025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-D,L-benzylsuccinic Acid
CAS 75043-31-1
Molekylær formel C11 H13 N O4
Mængde 25 mg
InChI formel InChI=1S/C11H13NO4/c12-9-3-1-7(2-4-9)5-8(11(15)16)6-10(13)14/h1-4,8H,5-6,12H2,(H,13,14)(H,15,16)
SMIL Nc1ccc(CC(CC(=O)O)C(=O)O)cc1
IUPAC navn 2-[(4-aminophenyl)methyl]butanedioic acid
Molekylvægt (g/mol) 223.23
Formel vægt 223.0845