Få mere at vide
4-Amino-6-(trifluoromethyl)benzene-1,3-disulphonamide, Mikromol™
Artikelnummer. 30049277
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049277 25 mg
30049622 100 mg
30460283 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30049277 Leverandør Mikromol Leverandørnr. MM0674.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Amino-6-(trifluoromethyl)benzene-1,3-disulphonamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-amino-6-(trifluoromethyl)benzene-1,3-disulfonamide
CAS 654-62-6
Analyt- eller komponentnavne 4-amino-6-(trifluoromethyl)benzene-1,3-disulfonamide
Forsendelsestilstand Room Temperature
Molekylær formel C7 H8 F3 N3 O4 S2
Certificeringer/compliance ISO 9001
Synonym Bendroflumethiazide Imp. A (EP), 2,4-Disulfamyl-5-Trifluoromethylaniline, 4-Amino-6-(trifluoromethyl)benzene-1,3-disulphonamide, Bendroflumethiazide Impurity A
InChI formel InChI=1S/C7H8F3N3O4S2/c8-7(9,10)3-1-4(11)6(19(13,16)17)2-5(3)18(12,14)15/h1-2H,11H2,(H2,12,14,15)(H2,13,16,17)
SMIL Nc1cc(c(cc1S(=O)(=O)N)S(=O)(=O)N)C(F)(F)F
IUPAC navn 4-amino-6-(trifluoromethyl)benzene-1,3-disulfonamide
Molekylvægt (g/mol) 319.28
Mængde 25 mg
Formel vægt 318.9908 g/mol