Få mere at vide
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid, Mikromol™
Artikelnummer. 17948964
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17948964 25 mg
17997314 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17948964 Leverandør Mikromol Leverandørnr. MM0793.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid
CAS 71675-87-1
Analyt- eller komponentnavne 4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C10 H13 N O5 S
Certificeringer/compliance ISO 17025
Synonym 4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid, Amisulpride Imp. E (EP), Amisulpride Impurity E
InChI formel InChI=1S/C10H13NO5S/c1-3-17(14,15)9-4-6(10(12)13)8(16-2)5-7(9)11/h4-5H,3,11H2,1-2H3,(H,12,13)
SMIL CCS(=O)(=O)c1cc(C(=O)O)c(OC)cc1N
IUPAC navn 4-amino-5-ethylsulfonyl-2-methoxybenzoic acid
Molekylvægt (g/mol) 259.28
Mængde 25 mg
Formel vægt 259.0514 g/mol