Få mere at vide
4-Amino-5-[(ethoxycarbonyl)amino]-1-methyl-d3 Uracil, TRC
Artikelnummer. 30309714
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30309714 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309714 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6091771MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-5-[(ethoxycarbonyl)amino]-1-methyl-d3 Uracil
CAS 1794768-19-6
Molekylær formel C8H9D3N4O4
Mængde 1 mg
Synonym (4-Amino-1,2,3,6-tetrahydro-1-methyl-d3-2,6-dioxo-5-pyrimidinyl)-carbamic Acid Ethyl Ester
InChI formel InChI=1S/C8H12N4O4/c1-3-16-8(15)10-4-5(9)11-7(14)12(2)6(4)13/h3,9H2,1-2H3,(H,10,15)(H,11,14)/i2D3
SMIL [2H]C([2H])([2H])N1C(=O)NC(=C(NC(=O)OCC)C1=O)N
IUPAC navn ethyl N-[6-amino-2,4-dioxo-3-(trideuteriomethyl)-1H-pyrimidin-5-yl]carbamate
Molekylvægt (g/mol) 231.22
Formel vægt 231.1047