Learn More
4-Amino-5-cyano-2-methylpyrimidine, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Amino-5-cyano-2-methylpyrimidine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water (6.4 g/L at 25°C). Soluble in dimethyl sulfoxide, ethanol and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4-Amino-5-cyano-2-methylpyrimidine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water (6.4 g/L at 25°C). Soluble in dimethyl sulfoxide, ethanol and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Color | Yellow |
Quantity | 1 g |
UN Number | UN3439 |
Solubility Information | Slightly soluble in water (6.4g/L at 25°C). Soluble in dimethyl sulfoxide,ethanol and methanol. |
Formula Weight | 134.14 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | 4-Amino-5-cyano-2-methylpyrimidine |