missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-cyano-2-methylpyrimidine, 95%, Thermo Scientific™

Catalog Number p-7053720
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
698-29-3
C6H6N4
134.14
MFCD00084875
YBPNIILOUYAGIF-UHFFFAOYSA-N
4-amino-5-cyano-2-methylpyrimidine, 5-pyrimidinecarbonitrile, 4-amino-2-methyl, 4-amino-2-methyl-5-pyrimidinecarbonitrile, pynitrile, pubchem6987, 2-methyl-4-amino-5-cyan-pyrimidin, 4-amino-5-cyano-2-methyl-pyrimidine, 2-methyl-4-amino-5-cyanopyrimidine, 4-amino-2-methyl-5-pyrimidine carbonitrile, 4-amino-2-methyl-pyrimidine-5-carbonitrile
69682
4-amino-2-methylpyrimidine-5-carbonitrile
CC1=NC=C(C#N)C(N)=N1
This item is not returnable. View return policy
698-29-3
C6H6N4
134.14
MFCD00084875
YBPNIILOUYAGIF-UHFFFAOYSA-N
4-amino-5-cyano-2-methylpyrimidine, 5-pyrimidinecarbonitrile, 4-amino-2-methyl, 4-amino-2-methyl-5-pyrimidinecarbonitrile, pynitrile, pubchem6987, 2-methyl-4-amino-5-cyan-pyrimidin, 4-amino-5-cyano-2-methyl-pyrimidine, 2-methyl-4-amino-5-cyanopyrimidine, 4-amino-2-methyl-5-pyrimidine carbonitrile, 4-amino-2-methyl-pyrimidine-5-carbonitrile
69682
4-amino-2-methylpyrimidine-5-carbonitrile
CC1=NC=C(C#N)C(N)=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Amino-5-cyano-2-methylpyrimidine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water (6.4 g/L at 25°C). Soluble in dimethyl sulfoxide, ethanol and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 250 mg
UN Number UN3439
Solubility Information Slightly soluble in water (6.4g/L at 25°C). Soluble in dimethyl sulfoxide,ethanol and methanol.
Formula Weight 134.14
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Amino-5-cyano-2-methylpyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'