Få mere at vide
4-Amino-5-chloro-2-ethoxybenzoic Acid-d5, TRC
Artikelnummer. 30314379
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30314379 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314379 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60358710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-5-chloro-2-ethoxybenzoic Acid-d5
CAS 1346600-23-4
Molekylær formel C9H5D5ClNO3
Mængde 10 mg
Synonym 2-Ethoxy-4-amino-5-chlorobenzoic Acid-d5, 4-Amino-5-chloro-2-ethoxybenzoic Acid-d5
InChI formel InChI=1S/C9H10ClNO3/c1-2-14-8-4-7(11)6(10)3-5(8)9(12)13/h3-4H,2,11H2,1H3,(H,12,13)/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Oc1cc(N)c(Cl)cc1C(=O)O
IUPAC navn 4-amino-5-chloro-2-(1,1,2,2,2-pentadeuterioethoxy)benzoic acid
Molekylvægt (g/mol) 220.66
Formel vægt 220.0663