missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-bromopyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055691
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1439-10-7
C4H4BrN3
174.001
MFCD00233962
IIFAONYUCDAVGA-UHFFFAOYSA-N
10261576
5-bromopyrimidin-4-amine
C1=C(C(=NC=N1)N)Br
This item is not returnable. View return policy
1439-10-7
C4H4BrN3
174.001
MFCD00233962
IIFAONYUCDAVGA-UHFFFAOYSA-N
10261576
5-bromopyrimidin-4-amine
C1=C(C(=NC=N1)N)Br

It is an inhibitor of thymidine phosphorylase, an enzyme known to be over-expressed in several tumor types and to play a role in metastasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an inhibitor of thymidine phosphorylase, an enzyme known to be over-expressed in several tumor types and to play a role in metastasis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 207°C to 211°C
Color Brown
Quantity 250 mg
Beilstein 112648
Synonym 4-amino-5-bromopyrimidine, 5-bromo-pyrimidin-4-ylamine, 4-pyrimidinamine, 5-bromo, 4-pyrimidinamine 5-bromo, 5-bromopyrimidine-4-ylamine, pubchem14978, acmc-209crn, 4-pyrimidinamine,5-bromo, 4-amino-5-bromo-pyrimidine, pyrimidine, 4-amino-5-bromo
Solubility Information Slightly soluble in water.
Formula Weight 174
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Amino-5-bromopyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'