missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Amino-5-bromopyrimidine, 98%

Brand:  Alfa Aesar™ H64335.MD

Additional Details : CAS Number : 1439-10-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15481499

  • 329.64 DKK / 250mg


In stock, ready to ship: 1
Add to basket

Description

Description

Applications
It is an inhibitor of thymidine phosphorylase, an enzyme known to be over-expressed in several tumor types and to play a role in metastasis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1439-10-7
174.001
IIFAONYUCDAVGA-UHFFFAOYSA-N
10261576
C1=C(C(=NC=N1)N)Br
C4H4BrN3
MFCD00233962
4-amino-5-bromopyrimidine, 5-bromo-pyrimidin-4-ylamine, 4-pyrimidinamine, 5-bromo, 4-pyrimidinamine 5-bromo, 5-bromopyrimidine-4-ylamine, pubchem14978, acmc-209crn, 4-pyrimidinamine,5-bromo, 4-amino-5-bromo-pyrimidine, pyrimidine, 4-amino-5-bromo
5-bromopyrimidin-4-amine

Specifications

4-Amino-5-bromopyrimidine
207°C to 211°C
1439-10-7
MFCD00233962
4-amino-5-bromopyrimidine, 5-bromo-pyrimidin-4-ylamine, 4-pyrimidinamine, 5-bromo, 4-pyrimidinamine 5-bromo, 5-bromopyrimidine-4-ylamine, pubchem14978, acmc-209crn, 4-pyrimidinamine,5-bromo, 4-amino-5-bromo-pyrimidine, pyrimidine, 4-amino-5-bromo
IIFAONYUCDAVGA-UHFFFAOYSA-N
5-bromopyrimidin-4-amine
10261576
Powder
Brown
250mg
C4H4BrN3
112648
Slightly soluble in water.
C1=C(C(=NC=N1)N)Br
174.001
174
98%
Videos
SDS Documents