missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Amino-5,6-dichloropyrimidine, 98%

Brand:  Alfa Aesar™ H64691.14

Additional Details : CAS Number : 310400-38-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15491999

  • 4120.00 DKK / 25g


Estimated Shipment
17-02-2021
Add to basket

Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

310400-38-5
163.989
ISTJUVRUUBNZNB-UHFFFAOYSA-N
24219262
C1=NC(=C(C(=N1)Cl)Cl)N
C4H3Cl2N3
MFCD09837293
4-amino-5,6-dichloropyrimidine, 5,6-dichloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro, 5,6-dichloro-4-pyrimidinamine, 5,6-dichloropyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro-9ci
5,6-dichloropyrimidin-4-amine

Specifications

4-Amino-5,6-dichloropyrimidine
25g
C4H3Cl2N3
8678505
Slightly soluble in water.
C1=NC(=C(C(=N1)Cl)Cl)N
163.989
163.99
98%
White
310400-38-5
MFCD09837293
4-amino-5,6-dichloropyrimidine, 5,6-dichloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro, 5,6-dichloro-4-pyrimidinamine, 5,6-dichloropyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro-9ci
ISTJUVRUUBNZNB-UHFFFAOYSA-N
5,6-dichloropyrimidin-4-amine
24219262
Solid
Videos
SDS Documents