missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Amino-3-nitrobenzamide, 97%

Catalog Number 15467597
Click to view available options
Quantity:
1g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 41263-65-4
Molecular Formula C7H7N3O3
MDL Number MFCD08729282
Quantity 1g
Synonym ambkkkkk269, benzamide, 4-amino-3-nitro
InChI Key YLKLDLKLEXDSJY-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(=O)N)[N+](=O)[O-])N
IUPAC Name 4-amino-3-nitrobenzamide
Molecular Weight (g/mol) 181.151
PubChem CID 12118056
Formula Weight 181.15
Percent Purity 97%
Chemical Name or Material 4-Amino-3-nitrobenzamide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'