Få mere at vide
4-Amino-3-methoxybenzeneethanamine-d4, TRC
Artikelnummer. 30325484
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30325484 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325484 Leverandør Toronto Research Chemicals Leverandørnr. TRCA612017250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-3-methoxybenzeneethanamine-d4
CAS 74719-63-4
Molekylær formel C16H15D4NO2
Mængde 250 mg
InChI formel InChI=1S/C16H19NO2/c1-18-16-11-13(9-10-17)7-8-15(16)19-12-14-5-3-2-4-6-14/h2-8,11H,9-10,12,17H2,1H3/i9D2,10D2
SMIL [2H]C([2H])(N)C([2H])([2H])c1ccc(OCc2ccccc2)c(OC)c1
IUPAC navn 1,1,2,2-tetradeuterio-2-(3-methoxy-4-phenylmethoxyphenyl)ethanamine
Molekylvægt (g/mol) 261.35
Formel vægt 261.1667