Få mere at vide
4-Amino-3-hydroxy Butanoic Acid Methyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Amino-3-hydroxy Butanoic Acid Methyl Ester Hydrochloride |
| CAS | 170726-96-2 |
| Molekylær formel | C5H12ClNO3 |
| Mængde | 500 mg |
| Synonym | Methyl 4-amino-3-hydroxybutyrate Hydrochloride, Methyl γ-Amino-β-hydroxybutyrate Hydrochloride, GABOB Methyl Ester Hydrochloride, γ-Amino-β-hydroxybutyric Acid Methyl Ester Hydrochloride |
| InChI formel | InChI=1S/C5H11NO3.ClH/c1-9-5(8)2-4(7)3-6;/h4,7H,2-3,6H2,1H3;1H |
| SMIL | NCC(O)CC(OC)=O.Cl |
| IUPAC navn | methyl 4-amino-3-hydroxybutanoate;hydrochloride |
| Molekylvægt (g/mol) | 169.61 |
| Formel vægt | 169.0506 |