Få mere at vide
4-Amino-3-butoxybenzoic Acid, Mikromol™
Artikelnummer. 30052248
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052248 25 mg
30050284 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052248 Leverandør Mikromol Leverandørnr. MM0543.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Amino-3-butoxybenzoic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-3-butoxybenzoic Acid
CAS 23442-22-0
Analyt- eller komponentnavne 4-Amino-3-butoxybenzoic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C11 H15 N O3
Certificeringer/compliance ISO 17025
Synonym Oxybuprocaine Hydrochloride Imp. B (EP), Oxybuprocaine Imp. B (EP), 4-Amino-3-butoxybenzoic Acid, Oxybuprocaine Hydrochloride Impurity B, Oxybuprocaine Impurity B
InChI formel InChI=1S/C11H15NO3/c1-2-3-6-15-10-7-8(11(13)14)4-5-9(10)12/h4-5,7H,2-3,6,12H2,1H3,(H,13,14)
SMIL CCCCOc1cc(ccc1N)C(=O)O
IUPAC navn 4-amino-3-butoxybenzoic acid
Molekylvægt (g/mol) 209.24
Mængde 25 mg
Formel vægt 209.1052 g/mol