Få mere at vide
4-Amino-2-sulfotoluene, TRC
Artikelnummer. 30137415
Skift visning
Mængde:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137415 25 g
30136851 10 g
30010468 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137415 Leverandør Toronto Research Chemicals Leverandørnr. TRCA29588025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 118-88-7
Molekylær formel C7 H9 NO3 S
Synonym 1-Amino-4-methylbenzene-3-sulfonic Acid∣2-Methyl-5-amino benzene sulfonic Acid∣3-Amino-6-methylbenzenesulfonic Acid∣3-Sulfo-4-methylaniline∣4-Amino-2-sulfotoluene∣4-Aminotoluene-2-sulfonic Acid∣4-Methyl-3-sulfoaniline∣4-Methylaniline-3-sulfonic Acid∣4-Toluidine-3-sulfonic Acid∣5-Amino-2-methyl-1-sulfobenzene∣5-Amino-o-toluenesulfonic Acid∣NSC 22958∣NSC 5586∣p-Toluidine-o-sulfonic Acid
Tekniske data
Kemisk navn eller materiale 5-Amino-2-methylbenzenesulfonic Acid
Mængde 25 g
Forsendelsestilstand Room Temperature
Formel vægt 187.0303
Molekylvægt (g/mol) 187.216, 187.21
InChI nøgle BRKFTWHPLMMNHF-UHFFFAOYSA-N
SMIL CC1=CC=C(N)C=C1S(O)(=O)=O
IUPAC navn 5-amino-2-methylbenzene-1-sulfonic acid