Få mere at vide
4-Amino-2-ethoxy-5-nitro-N-4-piperidinyl-benzamide Hydrobromide, TRC
Artikelnummer. 30134864
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134864 250 mg
30006630 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134864 Leverandør Toronto Research Chemicals Leverandørnr. TRCA608915250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 952310-00-8
Molekylær formel C14H21BrN4O4
Molekylvægt (g/mol) 389.24
Synonym 4-Amino-2-ethoxy-5-nitro-N-(piperidin-4-yl)benzamide Hydrobromide
IUPAC navn 4-amino-2-ethoxy-5-nitro-N-piperidin-4-ylbenzamide;hydrobromide
SMIL CCOC1=CC(=C(C=C1C(=O)NC2CCNCC2)[N+](=O)[O-])N.Br
Tekniske data
Kemisk navn eller materiale 4-Amino-2-ethoxy-5-nitro-N-4-piperidinyl-benzamide Hydrobromide
Analyt- eller komponentnavne 4-Amino-2-ethoxy-5-nitro-N-4-piperidinyl-benzamide Hydrobromide
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C14H20N4O4.BrH/c1-2-22-13-8-11(15)12(18(20)21)7-10(13)14(19)17-9-3-5-16-6-4-9;/h7-9,16H,2-6,15H2,1H3,(H,17,19);1H
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 250 mg
Formel vægt 388.0746