missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Amino-2-chloro-5-nitropyrimidine, 97%

Brand:  Alfa Aesar™ H64478.MD

Additional Details : CAS Number : 1920-66-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15491699

  • 232.69 DKK / 250mg


Estimated Shipment
26-10-2020
Add to basket

Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1920-66-7
174.544
RZGOEIWDMVQJBQ-UHFFFAOYSA-N
74716
C1=C(C(=NC(=N1)Cl)N)[N+](=O)[O-]
C4H3ClN4O2
MFCD00127771
4-amino-2-chloro-5-nitropyrimidine, 4-pyrimidinamine, 2-chloro-5-nitro, 2-chloro-4-amino-5-nitropyrimidine, 2-chloro-5-nitro-pyrimidin-4-ylamine, 2-chloro-5-nitro-4-pyrimidinamine, 4-pyrimidinamine,2-chloro-5-nitro, acmc-209x0o, 4-amino-5-nitro-2-chloropyrimidine, 2-chloro-5-nitropyrimidine-4-amine
2-chloro-5-nitropyrimidin-4-amine

Specifications

4-Amino-2-chloro-5-nitropyrimidine
250mg
C4H3ClN4O2
156953
Slightly soluble in water.
C1=C(C(=NC(=N1)Cl)N)[N+](=O)[O-]
174.544
174.54
97%
221°C to 226°C
1920-66-7
MFCD00127771
4-amino-2-chloro-5-nitropyrimidine, 4-pyrimidinamine, 2-chloro-5-nitro, 2-chloro-4-amino-5-nitropyrimidine, 2-chloro-5-nitro-pyrimidin-4-ylamine, 2-chloro-5-nitro-4-pyrimidinamine, 4-pyrimidinamine,2-chloro-5-nitro, acmc-209x0o, 4-amino-5-nitro-2-chloropyrimidine, 2-chloro-5-nitropyrimidine-4-amine
RZGOEIWDMVQJBQ-UHFFFAOYSA-N
2-chloro-5-nitropyrimidin-4-amine
74716
Solid
Videos
SDS Documents