Få mere at vide
4-Amino-2,2,6,6-tetramethylpiperidine-d17-1-oxyl, CDN
Artikelnummer. 30318725
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30318725 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318725 Leverandør CDN Leverandørnr. CDND27810.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl-D17
CAS 70588-04-4
Molekylær formel C9 D17 H2 N2 O
Mængde 0.01 g
Synonym 1-Piperidinyl-3,3,4,5,5-d5-oxy, 4-amino-2,2,6,6-tetra(methyl-d3)- (9 CI), 4-Amino-2,2,6,6-tetra(methyl-d3)-1-piperidinyl-3,3,4,5,5-d5-oxy, 4-Amino-2,2,6,6-tetramethylpiperidine-d17-1-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl-D17, 4-Aminotempo-d17, 4-NH2-TEMPO-d17, TAMINE-d17, Tempamine-d17, TEMPO-amine-d17
InChI formel InChI=1 S/C9H19N2O/c1-8(2)5-7(10)6-9(3,4)11(8)12/h7H,5-6,10H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7 D
SMIL [2 H]C([2 H])([2 H])C1(N([O])C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])(N)C1([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 3,3,4,5,5-pentadeuterio-1-λ1-oxidanyl-2,2,6,6-tetrakis(trideuteriomethyl)piperidin-4-amine
Molekylvægt (g/mol) 188.3647
Formel vægt 188.2564 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity