Få mere at vide
4-Amino-2,2,6,6-tetramethylpiperidine-d17; 1-15N-1-oxyl, TRC
Artikelnummer. 30324198
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324198 5 mg
30308944 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324198 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6302665MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl-D17-1-15N
CAS 97461-87-5
Molekylær formel C9 D17 H2 15N N O
Mængde 5 mg
Synonym 1-Piperidinyl-3,3,4,5,5-d5-1-15N-oxy, 4-amino-2,2,6,6-tetra(methyl-d3)- (9CI), 4-Amino-2,2,6,6-tetramethylpiperidine-d17, 1-15N-1-oxyl, 4-Amino-2,2,6,6-tetra(methyl-d3)-1-piperidinyl-3,3,4,5,5-d5-1-15N-oxy, 4-Amino-2,2,6,6-tetramethylpiperidine-d17-1-15N-1-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl-D17-1-15N, 4-Aminotempo-d17,15N, 4-NH2-TEMPO-d17,15N, TAMINE-d17,15N, Tempamine-d17,15N, TEMPO-amine-d17,15N
InChI formel InChI=1S/C9H19N2O/c1-8(2)5-7(10)6-9(3,4)11(8)12/h7H,5-6,10H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7D,11+1
SMIL [2H]C([2H])([2H])C1([15N]([O])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])(N)C1([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 3,3,4,5,5-pentadeuterio-1-λ1-oxidanyl-2,2,6,6-tetrakis(trideuteriomethyl)(115N)azinan-4-amine
Molekylvægt (g/mol) 189.3581
Formel vægt 189.2535