Få mere at vide
4-Amino-1-pentanol-d4, TRC
Artikelnummer. 30310522
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30310522 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310522 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61896810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-1-pentanol-d4
CAS 1216666-53-3
Molekylær formel C5H9D4NO
Mængde 10 mg
Synonym NSC 1095-d4
InChI formel InChI=1S/C5H13NO/c1-5(6)3-2-4-7/h5,7H,2-4,6H2,1H3/i2D2,4D2
SMIL [2H]C([2H])(O)C([2H])([2H])CC(C)N
IUPAC navn 4-amino-1,1,2,2-tetradeuteriopentan-1-ol
Molekylvægt (g/mol) 107.19
Formel vægt 107.1248