Få mere at vide
4-Amino-1-pentanol-d4 Hydrochloride Salt, TRC
Artikelnummer. 30324388
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30324388 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324388 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6189675MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-1-pentanol-d4 Hydrochloride Salt
CAS 1216414-18-4
Molekylær formel C5 D4 H9 N O . H Cl
Mængde 5 mg
InChI formel InChI=1S/C5H13NO.ClH/c1-5(6)3-2-4-7;/h5,7H,2-4,6H2,1H3;1H/i2D2,4D2;
SMIL Cl.[2H]C([2H])(O)C([2H])([2H])CC(C)N
IUPAC navn 4-amino-1,1,2,2-tetradeuteriopentan-1-ol;hydrochloride
Molekylvægt (g/mol) 143.648
Formel vægt 143.101