missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Amino-1-methylpyrimidin-2(1H)-one, TRC

Artikelnummer. 30346841
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30346841 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30346841 Leverandør Toronto Research Chemicals Leverandørnr. TRCA89821810MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Amino-1-methylpyrimidin-2(1H)-one
CAS 1122-47-0
Molekylær formel C5H7N3O
Mængde 10 mg
Synonym 1-Methylcytosine
InChI formel InChI=1S/C5H7N3O/c1-8-3-2-4(6)7-5(8)9/h2-3H,1H3,(H2,6,7,9)
SMIL CN1C=CC(=NC1=O)N
IUPAC navn 4-amino-1-methylpyrimidin-2-one
Molekylvægt (g/mol) 125.12
Formel vægt 125.0589
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.