Få mere at vide
4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide, Mikromol™
Artikelnummer. 30463344
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30463344 4 x 25 mg
30050851 25 mg
17912506 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30463344 Leverandør Mikromol Leverandørnr. MM0254.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
CAS 139756-02-8
Analyt- eller komponentnavne 4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
Forsendelsestilstand Room Temperature
Molekylær formel C8 H14 N4 O
Certificeringer/compliance ISO 17025
Synonym 4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
InChI formel InChI=1S/C8H14N4O/c1-3-4-5-6(9)7(8(10)13)12(2)11-5/h3-4,9H2,1-2H3,(H2,10,13)
Løsningstype Neat
SMIL CCCc1nn(C)c(C(=O)N)c1N
IUPAC navn 4-amino-2-methyl-5-propylpyrazole-3-carboxamide
Molekylvægt (g/mol) 182.22
Mængde 4 x 25 mg
Formel vægt 182.1168 g/mol