missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-(3-methyl-2-pyridyl)piperidine, 99%, Thermo Scientific™

Catalog Number 11494740
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
902837-09-6
C11H17N3
191.278
MFCD08060981
MEDSFHSPUSUMIL-UHFFFAOYSA-N
4-amino-1-3-methyl-2-pyridyl piperidine, 1-3-methylpyridin-2-yl piperidin-4-amine, acmc-20ampr, 1-3-methyl-2-pyridyl-4-piperidylamine, 1-3-methyl-2-pyridinyl-4-piperidinamine 3hcl
24208822
1-(3-methylpyridin-2-yl)piperidin-4-amine
CC1=C(N=CC=C1)N2CCC(CC2)N
This item is not returnable. View return policy
902837-09-6
C11H17N3
191.278
MFCD08060981
MEDSFHSPUSUMIL-UHFFFAOYSA-N
4-amino-1-3-methyl-2-pyridyl piperidine, 1-3-methylpyridin-2-yl piperidin-4-amine, acmc-20ampr, 1-3-methyl-2-pyridyl-4-piperidylamine, 1-3-methyl-2-pyridinyl-4-piperidinamine 3hcl
24208822
1-(3-methylpyridin-2-yl)piperidin-4-amine
CC1=C(N=CC=C1)N2CCC(CC2)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 191.28
Percent Purity 99%
Chemical Name or Material 4-Amino-1-(3-methyl-2-pyridyl)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'