Learn More
4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, 99%, BLD Pharm™
Product Code. 31334809
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31334809 5 g
31324010 100 g
31326391 10 g
31323711 1 g
31332242 25 g
31326039 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31334809 Supplier BLD Pharm Supplier No. BD1319265g
 Restricted Item
Forventet forsendelse: 25-11-2026
Specifications
CAS 95058-81-4
Molecular Formula C9H11F2N3O4
MDL Number MFCD00869720
Quantity 5 g
SMILES [C@H]2(N1C(N=C(N)C=C1)=O)C([C@H](O)[C@H](O2)CO)(F)F
Molecular Weight (g/mol) 263.20
Percent Purity 99%
Chemical Name or Material 4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one